Assay Details
Target Gene Details
Entrez Gene ID: | 53945 |
Gene Name: | solute carrier family 40 (iron-regulated transporter), member 1 |
Gene Aliases: |
Dusg, Fpn1, IREG1, MTP, MTP1, Ol5, Pcm, Slc11a3, Slc39a1 |
Location: |
Chr.1:45908070-45926535 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Slc40a1 | NM_016917.2 | NP_058613.2 | ||
| AF215637.1 | AAF80987.1 | |||
| AF226613.1 | AAF36696.1 | |||
| AF231120.1 | AAF44329.1 | |||
| AK008700.1 | BAB25840.1 | |||
| AK033987.1 | ||||
| AK144780.1 | BAE26063.1 | |||
| AK147137.1 | BAE27707.1 | |||
| AK159294.1 | BAE34969.1 | |||
| AK159855.1 | BAE35431.1 | |||
| AK167608.1 | BAE39662.1 | |||
| BB625854.1 | ||||
| BC003438.1 | AAH03438.1 | |||
| FJ207421.1 | ||||
| FJ207422.1 | ||||
| FJ207423.1 |
More Information
Set Membership: |
|

Genomic Map