Assay Details
Target Gene Details
Entrez Gene ID: | 69726 |
Gene Name: | SET and MYND domain containing 3 |
Gene Aliases: |
2410008A19Rik, Zmynd1 |
Location: |
Chr.1:178951960-179518052 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smyd3 | NM_027188.3 | NP_081464.1 | ||
| AK010447.1 | BAB26947.1 | |||
| AK043988.1 | ||||
| AK087728.1 | BAC39984.1 | |||
| AK163057.1 | BAE37175.1 | |||
| AK167875.1 | BAE39889.1 | |||
| BC052431.1 | AAH52431.1 | |||
| BC061485.1 | AAH61485.1 |
More Information
Set Membership: |
|

Genomic Map