Assay Details
Target Gene Details
Entrez Gene ID: | 20720 |
Gene Name: | serine (or cysteine) peptidase inhibitor, clade E, member 2 |
Gene Aliases: |
B230326M24Rik, PAI-1, PI-7, PI7, PN-1, Spi4 |
Location: |
Chr.1:79794319-79861175 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Serpine2 | NM_009255.4 | NP_033281.1 | ||
| AK045954.1 | BAC32546.1 | |||
| AK047856.1 | BAC33176.1 | |||
| AK051332.1 | BAC34605.1 | |||
| AK159687.1 | BAE35289.1 | |||
| BC010675.1 | AAH10675.1 | |||
| CT010311.1 | CAJ18519.1 | |||
| X70296.1 | CAA49777.1 |
More Information
Set Membership: |
|

Genomic Map