Assay Details
Target Gene Details
Entrez Gene ID: | 170755 |
Gene Name: | serum/glucocorticoid regulated kinase 3 |
Gene Aliases: |
2510015P22Rik, A330005P07Rik, Cisk, fy, fz |
Location: |
Chr.1:9798130-9902570 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Sgk3 | NM_001037759.1 | NP_001032848.1 | ||
| NM_133220.2 | NP_573483.1 | |||
| NM_177547.3 | NP_808215.2 | |||
| AF312007.1 | AAG34115.1 | |||
| AK028722.1 | ||||
| AK030314.1 | BAC26895.1 | |||
| AK031133.1 | BAC27269.1 | |||
| AK031328.1 | BAC27349.1 | |||
| AK143836.1 | ||||
| AK146561.1 | BAE27261.1 | |||
| AK151181.1 | BAE30182.1 | |||
| AK151793.1 | BAE30694.1 | |||
| AK151867.1 | BAE30755.1 | |||
| AK151996.1 | BAE30860.1 | |||
| AK166414.1 | ||||
| AK166531.1 | ||||
| AK171120.1 | BAE42262.1 | |||
| AK171186.1 | BAE42298.1 | |||
| AK171678.1 | ||||
| AK171959.1 | BAE42748.1 | |||
| AK172323.1 | BAE42945.1 | |||
| BB856423.1 | ||||
| BC018363.1 | AAH18363.1 |
More Information
Set Membership: |
|

Genomic Map