Assay Details
Target Gene Details
Entrez Gene ID: | 108989 |
Gene Name: | translocated promoter region, nuclear basket protein |
Gene Aliases: |
2610029M07Rik, C77892 |
Location: |
Chr.1:150392838-150449935 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tpr | NM_133780.3 | NP_598541.3 | ||
| AK076012.1 | BAC36119.1 | |||
| AK088078.1 | BAC40132.1 | |||
| AK132381.1 | BAE21135.1 | |||
| AK157716.1 | BAE34164.1 | |||
| AK157974.1 | BAE34294.1 | |||
| BC021320.1 | ||||
| BC108349.1 | AAI08350.1 | |||
| BC141406.1 | ||||
| BC141410.1 |
More Information
Set Membership: |
|

Genomic Map