Assay Details
Target Gene Details
Entrez Gene ID: | 22164 |
Gene Name: | tumor necrosis factor (ligand) superfamily, member 4 |
Gene Aliases: |
Ath-1, Ath1, CD134L, OX-40L, Ox40l, TXGP1, Txgp1l, gp34 |
Location: |
Chr.1:161395438-161418206 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tnfsf4 | NM_009452.2 | NP_033478.1 | ||
| FJ176943.1 | ||||
| U12763.1 | AAA21871.1 |
More Information
Set Membership: |
|

Genomic Map