Assay Details
Target Gene Details
Entrez Gene ID: | 30941 |
Gene Name: | ubiquitin specific peptidase 21 |
Gene Aliases: |
ESTM28, Usp16, Usp23, W53272 |
Location: |
Chr.1:171281949-171287961 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Usp21 | NM_013919.4 | NP_038947.2 | ||
| AF177759.1 | AAD54322.1 | |||
| AK011148.1 | BAB27431.1 | |||
| AK013252.1 | ||||
| BC021903.1 | AAH21903.1 | |||
| KF646669.1 |
More Information
Set Membership: |
|

Genomic Map