Assay Details
Target Gene Details
Entrez Gene ID: | 98403 |
Gene Name: | zinc finger protein 451 |
Gene Aliases: |
4930515K21Rik, 4933435G09Rik, AI596398, COASTER, Kiaa0576-hp, Znf451, mKIAA1702 |
Location: |
Chr.1:33761541-33815452 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Zfp451 | NM_001290699.1 | 3 | 388 | NP_001277628.1 |
| NM_001290700.1 | 3 | 388 | NP_001277629.1 | |
| NM_133817.3 | 3 | 388 | NP_598578.1 | |
| AK015797.1 | 3 | 363 | BAB29981.3 | |
| AK030088.1 | 3 | 221 | BAC26778.1 | |
| AK122541.1 | 3 | 213 | BAC65823.1 | |
| AK210231.1 | 2 | 135 | ||
| BC019546.1 | 3 | 224 | AAH19546.1 | |
| BC062154.1 | 3 | 178 | AAH62154.1 |
More Information
Set Membership: |
|

Genomic Map