Assay Details
Target Gene Details
Entrez Gene ID: | 211484 |
Gene Name: | testis specific 10 |
Gene Aliases: |
4933432N21Rik, AI843383, Gm217, Mtsga10 |
Location: |
Chr.1:37728144-37866739 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Tsga10 | NM_001290720.1 | NP_001277649.1 | ||
| NM_001290721.1 | NP_001277650.1 | |||
| NM_207228.3 | NP_997111.1 | |||
| NR_110976.1 | ||||
| AF530050.3 | AAN01136.3 | |||
| AK030254.1 | ||||
| AK149243.1 | 1 | 1163 | ||
| AK163138.1 | BAE37210.1 | |||
| AK164954.1 | ||||
| AK170053.1 | BAE41534.1 | |||
| AY618264.1 | AAT68665.1 | |||
| BC066782.1 | AAH66782.1 | |||
| CJ046680.1 |
More Information
Set Membership: |
|

Genomic Map