Assay Details
Target Gene Details
Entrez Gene ID: | 74104 |
Gene Name: | ATP-binding cassette, sub-family B (MDR/TAP), member 6 |
Gene Aliases: |
1200005B17Rik |
Location: |
Chr.1:75171640-75180283 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Abcb6 | NM_023732.2 | NP_076221.1 | ||
| AK004605.1 | BAB23404.1 | |||
| AK040323.1 | ||||
| AK151165.1 | BAE30168.1 | |||
| AK164003.1 | ||||
| AK168642.1 | BAE40501.1 | |||
| BC006634.1 | AAH06634.1 |
More Information
Set Membership: |
|

Genomic Map