Assay Details
Target Gene Details
Entrez Gene ID: | 226976 |
Gene Name: | KAT8 regulatory NSL complex subunit 3 |
Gene Aliases: |
4632411B12Rik, 4932435K23, AI431067, AI647574, C85723, Kiaa1310, mKIAA1310 |
Location: |
Chr.1:36329947-36369194 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kansl3 | NM_172652.3 | 3 | 390 | NP_766240.1 |
| AK030079.1 | 3 | 391 | BAC26771.1 | |
| AK129329.1 | BAC98139.1 | |||
| BC132293.1 | 2 | 293 |

Genomic Map