Assay Details
Target Gene Details
Entrez Gene ID: | 76187 |
Gene Name: | alcohol dehydrogenase, iron containing, 1 |
Gene Aliases: |
6330565B14Rik, AI043035, Adh8 |
Location: |
Chr.1:9547969-9577968 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Adhfe1 | NM_175236.4 | NP_780445.1 | ||
| NR_027664.1 | ||||
| AK018241.1 | ||||
| AK038853.1 | BAC30151.1 | |||
| AK046780.1 | ||||
| AK050178.1 | BAC34108.1 | |||
| BC026584.1 | AAH26584.2 | |||
| CA467848.1 |
More Information
Set Membership: |
|

Genomic Map