Assay Details
Target Gene Details
Entrez Gene ID: | 347722 |
Gene Name: | ArfGAP with GTPase domain, ankyrin repeat and PH domain 1 |
Gene Aliases: |
Centg2, Ggap1, R75364, mKIAA1099 |
Location: |
Chr.1:89454811-89895282 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Agap1 | NM_001037136.1 | NP_001032213.1 | ||
| NM_178119.3 | NP_835220.1 | |||
| AK046523.1 | BAC32770.1 | |||
| AK129289.2 | BAC98099.2 | |||
| AK147503.1 | BAE27957.1 | |||
| AK147521.1 | BAE27969.1 |

Genomic Map