Assay Details
Target Gene Details
Entrez Gene ID: | 227290 |
Gene Name: | angio-associated migratory protein |
Gene Aliases: |
AU040907, Aamp-rs |
Location: |
Chr.1:74279840-74284738 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Aamp | NM_001190444.1 | 3 | 451 | NP_001177373.1 |
| NM_146110.3 | 3 | 448 | NP_666222.2 | |
| AK146470.1 | 3 | 391 | BAE27196.1 | |
| AK151820.1 | 3 | 360 | BAE30717.1 | |
| AK162750.1 | 3 | 376 | BAE37048.1 | |
| AK167621.1 | 3 | 372 | BAE39673.1 | |
| AK180202.1 | 2 | 92 | ||
| AK183212.1 | 2 | 93 | ||
| AK189765.1 | 2 | 92 | ||
| AK200115.1 | 2 | 92 | ||
| AK212494.1 | 2 | 92 | ||
| BC031795.1 | 2 | 140 | AAH31795.1 |

Genomic Map