Assay Details
Target Gene Details
Entrez Gene ID: | 226747 |
Gene Name: | AT hook containing transcription factor 1 |
Gene Aliases: |
6230412P20Rik, AV011447, Elys |
Location: |
Chr.1:179744894-179804015 on Build GRCm38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ahctf1 | NM_026375.2 | 24 | 3695 | NP_080651.2 |
| AB059278.1 | 24 | 3351 | BAB78517.1 | |
| BC138343.1 | 24 | 3338 | ||
| BC138344.1 | 24 | 3338 |
More Information
Set Membership: |
|

Genomic Map