Assay Details
Target Gene Details
Entrez Gene ID: | 71724 |
Gene Name: | aldehyde oxidase 3 |
Gene Aliases: |
1200011D03Rik, AI326299, Aoh1, Aoh2 |
Location: |
Chr.1:58113136-58200452 on Build GRCm38 |
Assay Gene Location: | Within Exon 25 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Aox3 | NM_023617.2 | 25 | 2818 | NP_076106.2 |
AF172276.2 | 25 | 2894 | AAD51028.2 | |
AK004700.1 | 14 | 1614 | BAB23485.1 | |
AK030109.1 | 21 | 2358 | ||
AK149434.1 | 25 | 2779 | ||
AY347569.1 | 25 | 2695 | AAQ24537.1 | |
AY347570.1 | 25 | 2695 | AAQ24538.1 | |
BC138876.1 | 25 | 2797 |
