Assay Details
Target Gene Details
Entrez Gene ID: | 108664 |
Gene Name: | ATPase, H+ transporting, lysosomal V1 subunit H |
Gene Aliases: |
0710001F19Rik, AU022349, CGI-11, SFD, SFDalpha, SFDbeta, VMA13 |
Location: |
Chr.1:5082978-5162549 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Atp6v1h | NM_133826.4 | 10 | 1139 | NP_598587.2 |
| AK034165.1 | 8 | 3298 | ||
| AK078767.1 | 10 | 1140 | BAC37382.1 | |
| AK152147.1 | 10 | 1161 | ||
| BC009154.1 | 10 | 1140 | AAH09154.1 |
More Information
Set Membership: |
|

Genomic Map