Assay Details
Target Gene Details
Entrez Gene ID: | 280645 |
Gene Name: | beta-1,3-glucuronyltransferase 2 (glucuronosyltransferase S) |
Gene Aliases: |
GlcAT-S, Glcats |
Location: |
Chr.1:23761926-23847865 on Build GRCm38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| B3gat2 | NM_172124.2 | 3 | 1520 | NP_742122.2 |
| AB055902.1 | 3 | 1466 | BAC20343.1 | |
| AK052640.1 | 3 | 1520 | BAC35075.1 | |
| AK136293.1 | 3 | 441 | BAE22919.1 | |
| BC056368.1 | 3 | 1412 | AAH56368.1 | |
| BC058082.1 | 3 | 1412 | AAH58082.1 |
Target Gene Details
Entrez Gene ID: | 98366 |
Gene Name: | small ArfGAP 1 |
Gene Aliases: |
1700056O10Rik, 4921514B13Rik, 4921525H11Rik, AI462175 |
Location: |
Chr.1:23844825-23922450 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Smap1 | NR_110972.1 | 11 | 1766 | |
| AK132868.1 | 11 | 1767 | ||
| BE631392.1 | 1 | 397 |
More Information
Set Membership: |
|

Genomic Map