Assay Details
Target Gene Details
Entrez Gene ID: | 213539 |
Gene Name: | BCL2-associated athanogene 2 |
Gene Aliases: |
2610042A13Rik, BC016230 |
Location: |
Chr.1:33745484-33757750 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Bag2 | NM_145392.2 | NP_663367.1 | ||
| AK083600.1 | BAC38967.1 | |||
| AK144720.1 | BAE26031.1 | |||
| BC016230.1 | AAH16230.1 | |||
| BC028944.1 | AAH28944.1 | |||
| BY014303.1 |
More Information
Set Membership: |
|

Genomic Map