Assay Details
Target Gene Details
Entrez Gene ID: | 226499 |
Gene Name: | cDNA sequence BC003331 |
Gene Aliases: |
1810053E15Rik, Odr4, odr-4 |
Location: |
Chr.1:150361311-150393055 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BC003331 | NM_001077237.1 | NP_001070705.1 | ||
| NM_145511.2 | NP_663486.2 | |||
| AK152022.1 | BAE30884.1 | |||
| AK152955.1 | ||||
| BC003331.1 | ||||
| DQ077804.2 |

Genomic Map