Assay Details
Target Gene Details
Entrez Gene ID: | 226499 |
Gene Name: | cDNA sequence BC003331 |
Gene Aliases: |
1810053E15Rik, Odr4, odr-4 |
Location: |
Chr.1:150361311-150393055 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BC003331 | NM_001077237.1 | 7 | 933 | NP_001070705.1 |
| NM_145511.2 | 7 | 933 | NP_663486.2 | |
| AK152022.1 | 5 | 449 | BAE30884.1 | |
| AK152955.1 | 5 | 449 | ||
| BC003331.1 | 6 | 588 | ||
| CB235715.1 | 7 | 631 | ||
| CX243768.1 | 3 | 181 | ||
| DQ077804.2 | 6 | 547 |

Genomic Map