Assay Details
Target Gene Details
Entrez Gene ID: | 227485 |
Gene Name: | cadherin 19, type 2 |
Gene Aliases: |
- |
Location: |
Chr.1:110888002-110977593 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdh19 | NM_001081386.1 | NP_001074855.1 | ||
| AK148396.1 | BAE28530.1 | |||
| BC157927.1 |
More Information
Set Membership: |
|

Genomic Map