Assay Details
Target Gene Details
Entrez Gene ID: | 13211 |
Gene Name: | DEAH (Asp-Glu-Ala-His) box polypeptide 9 |
Gene Aliases: |
AI326842, Ddx9, HEL-5, NDHII, RHA, mHEL-5 |
Location: |
Chr.1:153455756-153487686 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dhx9 | NM_007842.2 | 8 | 907 | NP_031868.2 |
| AK013423.1 | BAB28848.1 | |||
| AK135603.1 | 3 | 221 | ||
| AK141824.1 | 8 | 902 | BAE24846.1 | |
| AK148423.1 | 8 | 903 | ||
| AY512925.1 | 4 | 363 | AAR87796.1 | |
| BC089159.1 | 8 | 901 | AAH89159.1 | |
| U91922.1 | 8 | 864 | AAC05725.1 |
More Information
Set Membership: |
|

Genomic Map