Assay Details
Target Gene Details
Entrez Gene ID: | 627872 |
Gene Name: | dynein, axonemal, heavy chain 7A |
Gene Aliases: |
Dnahc7, Dnahc7a, Gm992 |
Location: |
Chr.1:53396494-53708118 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dnah7a | NM_001252070.1 | NP_001238999.1 |
More Information
Set Membership: |
|

Genomic Map