Assay Details
Target Gene Details
Entrez Gene ID: | 12767 |
Gene Name: | chemokine (C-X-C motif) receptor 4 |
Gene Aliases: |
CD184, Cmkar4, LESTR, PB-CKR, PBSF/SDF-1, Sdf1r, b2b220Clo |
Location: |
Chr.1:128588199-128592315 on Build GRCm38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cxcr4 | NM_009911.3 | 1 | 67 | NP_034041.2 |
| AB000803.1 | BAA19187.1 | |||
| BB872868.1 | 1 | 68 | ||
| BC031665.1 | AAH31665.1 | |||
| BC098322.1 | AAH98322.1 | |||
| U59760.1 | AAB07725.1 | |||
| X99582.1 | 1 | 67 | CAA67894.1 | |
| Z80111.1 | 1 | 61 | CAB02201.1 |

Genomic Map