Assay Details
Target Gene Details
Entrez Gene ID: | 14262 |
Gene Name: | flavin containing monooxygenase 3 |
Gene Aliases: |
AW111792 |
Location: |
Chr.1:162953800-162984491 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fmo3 | NM_008030.1 | NP_032056.1 | ||
| AK050123.1 | BAC34077.1 | |||
| AK149440.1 | BAE28877.1 | |||
| U87147.1 | AAB47541.1 |
More Information
Set Membership: |
|

Genomic Map