Assay Details
Target Gene Details
Entrez Gene ID: | 226564 |
Gene Name: | flavin containing monooxygenase 4 |
Gene Aliases: |
D1Ertd532e |
Location: |
Chr.1:162793188-162815869 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fmo4 | NM_144878.1 | NP_659127.1 | ||
| AF461145.1 | AAL66366.1 | |||
| AK146562.1 | ||||
| BC106858.1 | AAI06859.1 | |||
| BC106859.1 | AAI06860.1 |
More Information
Set Membership: |
|

Genomic Map