Assay Details
Target Gene Details
Entrez Gene ID: | 269132 |
Gene Name: | collagen beta(1-O)galactosyltransferase 2 |
Gene Aliases: |
AI427652, D730024P12, Glt25d2 |
Location: |
Chr.1:152399867-152510695 on Build GRCm38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Colgalt2 | NM_177756.4 | 2 | 580 | NP_808424.3 |
| AK052838.1 | 2 | 581 | BAC35169.1 | |
| AK076222.1 | 2 | 322 | ||
| AK085716.1 | 2 | 618 | ||
| AK134084.1 | 2 | 546 | ||
| BC068118.1 | 2 | 526 | AAH68118.1 |
More Information
Set Membership: |
|

Genomic Map