Assay Details
Target Gene Details
Entrez Gene ID: | 214469 |
Gene Name: | family with sequence similarity 168, member B |
Gene Aliases: |
mKIAA4042 |
Location: |
Chr.1:34813218-34843050 on Build GRCm38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fam168b | NM_001160235.1 | 7 | 815 | NP_001153707.1 |
| NM_001160236.1 | 6 | 800 | NP_001153708.1 | |
| NM_174997.2 | 6 | 779 | NP_778162.1 | |
| AK144298.1 | 6 | 679 | BAE25820.1 | |
| AK198592.1 | ||||
| AK220453.1 | 6 | 779 | BAD90278.1 | |
| BC040511.1 | 6 | 756 | AAH40511.1 | |
| BC043098.1 | 6 | 703 | AAH43098.1 | |
| BC059247.1 | 6 | 756 | AAH59247.1 | |
| CA328263.1 | 5 | 548 | ||
| CF538379.1 |
More Information
Set Membership: |
|

Genomic Map