Assay Details
Target Gene Details
Entrez Gene ID: | 98752 |
Gene Name: | Fc receptor-like A |
Gene Aliases: |
BB219290, FCRL, FCRL1, FREB, Fcrlm1, Fcrx, Freb1, mFREB, mFcrX |
Location: |
Chr.1:170917594-170927590 on Build GRCm38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Fcrla | NM_001160215.1 | NP_001153687.1 | ||
| NM_145141.2 | NP_660123.1 | |||
| NR_027666.1 | ||||
| AF329487.1 | AAL23897.1 | |||
| AF426462.1 | AAL58112.1 | |||
| AF531424.1 | AAM97592.1 | |||
| AK041572.1 | BAC30990.1 | |||
| AK172370.1 | BAE42968.1 | |||
| BB637253.1 | ||||
| BC064708.1 | AAH64708.1 | |||
| CJ080700.1 |
More Information
Set Membership: |
|

Genomic Map