Assay Details
Target Gene Details
Entrez Gene ID: | 98488 |
Gene Name: | general transcription factor IIIC, polypeptide 3 |
Gene Aliases: |
AL022818, TF3C-gamma, TFIIIC102 |
Location: |
Chr.1:54397581-54438971 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Gtf3c3 | NM_001033194.2 | NP_001028366.1 | ||
| AK160997.1 | ||||
| AK165830.1 | BAE38400.1 | |||
| AK179332.1 | ||||
| BC141157.1 |
More Information
Set Membership: |
|

Genomic Map