Assay Details
Target Gene Details
Entrez Gene ID: | 226438 |
Gene Name: | immunoglobulin-like and fibronectin type III domain containing 1 |
Gene Aliases: |
9830123M21Rik |
Location: |
Chr.1:135953205-136040842 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 23 - Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Igfn1 | NM_177642.3 | NP_808310.2 | ||
| AK036509.1 | BAC29455.1 | |||
| BC107027.1 | AAI07028.1 | |||
| BC107028.2 | AAI07029.1 | |||
| EU429481.1 | ||||
| EU429482.1 | ||||
| EU429483.2 | ||||
| GU290042.1 |

Genomic Map