Assay Details
Target Gene Details
Entrez Gene ID: | 74106 |
Gene Name: | DDB1 and CUL4 associated factor 6 |
Gene Aliases: |
1200006M05Rik, Iqwd1, PC326 |
Location: |
Chr.1:165329500-165460553 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 12 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Dcaf6 | NM_028759.1 | NP_083035.1 | ||
| AK004618.1 | BAB23414.1 | |||
| AK077238.1 | BAC36701.1 | |||
| AK161893.1 |
More Information
Set Membership: |
|

Genomic Map