Assay Details
Target Gene Details
Entrez Gene ID: | 227195 |
Gene Name: | INO80 complex subunit D |
Gene Aliases: |
7330405I11, A430093A21Rik, BC010584, OTTMUSG00000002594 |
Location: |
Chr.1:63047801-63114267 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
Ino80d | NM_001081436.2 | NP_001074905.1 | ||
NM_001114609.1 | NP_001108081.1 | |||
AK049468.1 | BAC33763.1 | |||
BC080705.1 | AAH80705.1 | |||
BC150989.1 |
