Assay Details
Target Gene Details
Entrez Gene ID: | 226844 |
Gene Name: | major facilitator superfamily domain containing 7B |
Gene Aliases: |
9630055N22Rik, FLVCR, Flvcr1 |
Location: |
Chr.1:191005832-191026215 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mfsd7b | NM_001081259.1 | NP_001074728.1 | ||
| AK036315.1 | ||||
| AK165872.1 | ||||
| BC024751.1 | ||||
| BC157992.1 |
More Information
Set Membership: |
|

Genomic Map