Assay Details
Target Gene Details
Entrez Gene ID: | 16560 |
Gene Name: | kinesin family member 1A |
Gene Aliases: |
ATSV, C630002N23Rik, Gm1626, Kns1 |
Location: |
Chr.1:93015455-93101890 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 48 - Exon 49 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Kif1a | NM_001110315.2 | NP_001103785.1 | ||
| NM_001294149.1 | NP_001281078.1 | |||
| NM_001294150.1 | NP_001281079.1 | |||
| NM_008440.4 | NP_032466.2 | |||
| AK147640.1 | BAE28041.1 | |||
| AK220487.1 | BAD90508.1 | |||
| AY426748.1 | AAR04821.1 | |||
| BC062891.1 | AAH62891.1 | |||
| D29951.1 | BAA06221.1 |
More Information
Set Membership: |
|

Genomic Map