Assay Details
Target Gene Details
Entrez Gene ID: | 381338 |
Gene Name: | LON peptidase N-terminal domain and ring finger 2 |
Gene Aliases: |
2900060P06Rik, AI851349 |
Location: |
Chr.1:38793669-38836719 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lonrf2 | NM_001029878.1 | NP_001025049.1 | ||
| BC139127.1 | ||||
| BC139130.1 |
More Information
Set Membership: |
|

Genomic Map