Assay Details
Target Gene Details
Entrez Gene ID: | 98558 |
Gene Name: | maelstrom homolog (Drosophila) |
Gene Aliases: |
4933405K18Rik, AU019877 |
Location: |
Chr.1:166201385-166238744 on Build GRCm38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mael | NM_175296.4 | 6 | 668 | NP_780505.1 |
| AK077103.1 | 6 | 665 | BAC36612.1 | |
| AK165801.1 | 6 | 665 | BAE38386.1 | |
| BC050800.1 | 6 | 668 | AAH50800.1 |

Genomic Map