Assay Details
Target Gene Details
Entrez Gene ID: | 17215 |
Gene Name: | minichromosome maintenance deficient 3 (S. cerevisiae) |
Gene Aliases: |
AL033361, C80350, Mcmd, P1, p1.m |
Location: |
Chr.1:20803014-20820213 on Build GRCm38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Mcm3 | NM_008563.2 | 10 | 1464 | NP_032589.1 |
| AK088142.1 | 10 | 1513 | ||
| AK133850.1 | 10 | 1529 | BAE21885.1 | |
| AK145563.1 | 10 | 1563 | BAE26512.1 | |
| AK146497.1 | 10 | 1477 | BAE27213.1 | |
| AK147065.1 | 10 | 1479 | BAE27649.1 | |
| AK148006.1 | 10 | 1509 | BAE28283.1 | |
| AK178723.1 | ||||
| AK188323.1 | ||||
| BC031700.1 | 10 | 1464 | AAH31700.1 | |
| X62154.1 | 9 | 1408 | CAA44079.1 |
More Information
Set Membership: |
|

Genomic Map