Assay Details
Target Gene Details
Entrez Gene ID: | 329252 |
Gene Name: | leucine-rich repeat-containing G protein-coupled receptor 6 |
Gene Aliases: |
A530037C04Rik, D830026M09 |
Location: |
Chr.1:134983300-135105374 on Build GRCm38 |
Assay Gene Location: | Within Exon 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Lgr6 | NM_001033409.3 | 16 | 1621 | NP_001028581.1 |
| AK052873.1 | 11 | 821 | ||
| AK085901.1 | 16 | 1580 | ||
| AK137380.1 | 16 | 1622 | BAE23335.1 | |
| BC026896.1 | AAH26896.1 | |||
| BC141210.1 | 16 | 1573 |

Genomic Map