Assay Details
Target Gene Details
Entrez Gene ID: | 100380944 |
Gene Name: | INO80 complex subunit D, opposite strand |
Gene Aliases: |
Gm11602, OTTMUSG00000002611 |
Location: |
Chr.1:63114471-63158295 on Build GRCm38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ino80dos | NR_045914.1 | |||
| NR_045915.1 | ||||
| AK017924.1 |
Target Gene Details
Entrez Gene ID: | 227197 |
Gene Name: | NADH dehydrogenase (ubiquinone) Fe-S protein 1 |
Gene Aliases: |
5830412M15Rik, 9930026A05Rik |
Location: |
Chr.1:63143592-63176822 on Build GRCm38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ndufs1 | NM_001160038.1 | 17 | 2339 | NP_001153510.1 |
| NM_001160039.1 | 17 | 2181 | NP_001153511.1 | |
| NM_001160040.1 | 16 | 2235 | NP_001153512.1 | |
| NM_145518.2 | 16 | 2077 | NP_663493.2 | |
| AK034597.1 | 16 | 2146 | BAE20494.1 | |
| AK036926.1 | 16 | 1942 | BAC29641.1 | |
| AK142711.1 | 16 | 1957 | BAE25170.1 | |
| AK146814.1 | 16 | 2095 | BAE27454.1 | |
| AK167705.1 | 16 | 1925 | BAE39749.1 | |
| AK196215.1 | 3 | 324 | ||
| AK219412.1 | 3 | 324 | ||
| BC006660.1 | 16 | 1964 | AAH06660.1 | |
| BC015300.1 | 16 | 1897 | AAH15300.1 |
More Information
Set Membership: |
|

Genomic Map