Assay Details
Target Gene Details
Entrez Gene ID: | 66208 |
Gene Name: | neuron derived neurotrophic factor |
Gene Aliases: |
1110060M21Rik, SCIRP10, Spuf |
Location: |
Chr.1:191306797-191318118 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Nenf | NM_025424.2 | NP_079700.1 | ||
| AB126218.1 | BAD72062.1 | |||
| AK002413.1 | BAB22081.1 | |||
| AK004329.1 | BAB23264.1 | |||
| AK009304.1 | BAB26205.1 | |||
| AK020403.1 | BAB32092.1 | |||
| AK155319.1 | BAE33187.1 | |||
| BC048464.1 | AAH48464.1 |
More Information
Set Membership: |
|

Genomic Map