Assay Details
Target Gene Details
Entrez Gene ID: | 27392 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis, class N |
Gene Aliases: |
Gm20308, PIG-N |
Location: |
Chr.1:105518422-105663722 on Build GRCm38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pign | NM_013784.3 | 8 | 1030 | NP_038812.2 |
| AB030279.1 | 8 | 1016 | BAA82619.1 | |
| AB030280.1 | 8 | 1016 | BAA82620.1 | |
| AK014165.1 | 8 | 1002 | ||
| AK165971.1 | 7 | 910 | ||
| BC021148.1 | 8 | 1014 | AAH21148.1 |
More Information
Set Membership: |
|

Genomic Map