Assay Details
Target Gene Details
Entrez Gene ID: | 23850 |
Gene Name: | pappalysin 2 |
Gene Aliases: |
PAPP-A2, PLAC3, Pappe |
Location: |
Chr.1:158711727-158980523 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pappa2 | NM_001085376.2 | NP_001078845.1 | ||
| AK084006.1 | BAC39093.1 | |||
| BC094560.1 | AAH94560.1 |

Genomic Map