Assay Details
Target Gene Details
Entrez Gene ID: | 18143 |
Gene Name: | neuronal PAS domain protein 2 |
Gene Aliases: |
MOP4, bHLHe9 |
Location: |
Chr.1:39193956-39363240 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Npas2 | NM_008719.2 | NP_032745.2 | ||
| BC109166.2 | AAI09167.1 | |||
| U77969.1 | AAB47249.1 |

Genomic Map