Assay Details
Target Gene Details
Entrez Gene ID: | 271697 |
Gene Name: | cyclin-dependent kinase 15 |
Gene Aliases: |
Als2cr7, Pftk2 |
Location: |
Chr.1:59256889-59352612 on Build GRCm38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Cdk15 | NM_001033373.2 | 12 | 1184 | NP_001028545.2 |
| AK042881.1 | BAE20636.1 | |||
| AK181908.1 | 2 | 106 | ||
| AK197819.1 | 1 | 73 | ||
| AK211120.1 | 5 | 330 | ||
| AK219417.1 | 2 | 106 | ||
| BC147314.1 | 12 | 1184 | ||
| BC147315.1 | 12 | 1184 | ||
| BC147758.1 | 12 | 1184 | ||
| BC147763.1 | 12 | 1184 | ||
| BC172036.1 | 12 | 1181 |
More Information
Set Membership: |
|

Genomic Map