Assay Details
Target Gene Details
Entrez Gene ID: | 56695 |
Gene Name: | paroxysmal nonkinesiogenic dyskinesia |
Gene Aliases: |
2210013N15Rik, 2810403H05Rik, AI854243, Brp17, MNCb-5687, MR-1, Tahccp2 |
Location: |
Chr.1:74284597-74353692 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 5 - Intron 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Pnkd | NM_001039509.1 | NP_001034598.1 | ||
| NM_019999.2 | NP_064383.1 | |||
| AB041609.1 | BAA95092.1 | |||
| AK041238.1 | BAC30873.1 | |||
| AK173125.1 | BAD32403.1 | |||
| BC058945.1 | AAH58945.1 |

Genomic Map