Assay Details
Target Gene Details
Entrez Gene ID: | 68507 |
Gene Name: | protein tyrosine phosphatase, receptor type, f polypeptide (PTPRF), interacting protein (liprin), alpha 4 |
Gene Aliases: |
1110008G13Rik, AI448359, AI852265, C81506 |
Location: |
Chr.1:134296783-134344750 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 27 - Exon 28 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Ppfia4 | NM_001144855.1 | NP_001138327.1 | ||
| AK003571.1 | ||||
| AK078263.1 | ||||
| AK170782.1 | ||||
| AK173053.1 | BAD32331.1 | |||
| EU568872.1 |

Genomic Map