Assay Details
Target Gene Details
Entrez Gene ID: | 18802 |
Gene Name: | phospholipase C, delta 4 |
Gene Aliases: |
4921507K24Rik |
Location: |
Chr.1:74541972-74565977 on Build GRCm38 |
Assay Gene Location: | Overlaps Exon 13 - Intron 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Plcd4 | NM_001081456.1 | NP_001074925.1 | ||
| NM_148937.2 | NP_683739.2 | |||
| AK029561.1 | ||||
| AK140231.1 | BAE24292.1 | |||
| AY033991.1 | AAK61537.1 | |||
| BC066156.1 | AAH66156.1 |
More Information
Set Membership: |
|

Genomic Map