Assay Details
Target Gene Details
Entrez Gene ID: | 623503 |
Gene Name: | prolactin releasing hormone |
Gene Aliases: |
EG623503 |
Location: |
Chr.1:90951256-90954369 on Build GRCm38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| Prlh | NM_001101647.1 | NP_001095117.1 | ||
| FJ424613.1 |

Genomic Map